Sign In to View Organizational & Contract Pricing.
About This Item
eCl@ss:
32160702
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
74.5.2, monoclonal
Application:
ELISA, FACS, IF, WB
Citations:
4
biological source
mouse
conjugate
unconjugated
antibody form
ascites fluid
antibody product type
primary antibodies
clone
74.5.2, monoclonal
species reactivity
human
manufacturer/tradename
Upstate®
technique(s)
ELISA: suitable, flow cytometry: suitable, immunofluorescence: suitable, western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... C1QBP(708)
General description
33kDa
Immunogen
GST fusion protein corresponding to residues 74-282 of the mature form of human gC1qR. The epitope has been mapped to residues 204-218
Application
Anti-gC1qR Antibody, clone 74.5.2 detects level of gC1qR & has been published & validated for use in ELISA, FC, IF & WB.
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Immunoglobulins & Immunology
Biochem/physiol Actions
mature form and the truncated form of gC1qR
Physical form
Ascites
mouse ascites with 30% glycerol
Preparation Note
2 years at -20°C
Analysis Note
routinely evaluated by immunoblot on RIPA lysates from Raji cells
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.