Skip to Content
Merck

Skip To

05-23-0751

α-Melanocyte-Stimulating Hormone

Synthetic α-melanocyte-stimulating hormone.

Synonym(s):

α-Melanocyte-Stimulating Hormone, α-MSH, Melanotropin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
CZK 2,630.00

About This Item

Empirical Formula (Hill Notation):
C77H109N21O19S
CAS Number:
Molecular Weight:
1664.88
UNSPSC Code:
12352202
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
lyophilized
Storage condition:
OK to freeze

CZK 2,630.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

non-sterile

assay

≥95% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)CO)Cc6ccc(cc6)O)CO)C(=O)N[C@@H](CCC(=O)O)C(=O)NC(Cc5[nH]cnc5)C(=O)N[C@@H](Cc4ccccc4)C(=O)N[C@@H](CCCN=C(N)N)C(=O)N[C@@H](Cc2c3c([nH]c2)cccc3)C(=O)NCC(=O)N[C@@H](CCCCN)C(=O)N1[C@@H](CCC1)C(=O)N[C@@H]

InChI

1S/C77H109N21O19S/c1-42(2)64(65(79)106)97-75(116)61-20-13-30-98(61)76(117)54(18-10-11-28-78)88-62(103)38-85-66(107)57(34-46-36-84-50-17-9-8-16-49(46)50)94-67(108)51(19-12-29-83-77(80)81)89-70(111)55(32-44-14-6-5-7-15-44)92-72(113)58(35-47-37-82-41-86-47)95-68(109)52(25-26-63(104)105)90-69(110)53(27-31-118-4)91-74(115)60(40-100)96-71(112)56(33-45-21-23-48(102)24-22-45)93-73(114)59(39-99)87-43(3)101/h5-9,14-17,21-24,36-37,41-42,51-61,64,84,99-100,102H,10-13,18-20,25-35,38-40,78H2,1-4H3,(H2,79,106)(H,82,86)(H,85,107)(H,87,101)(H,88,103)(H,89,111)(H,90,110)(H,91,115)(H,92,113)(H,93,114)(H,94,108)(H,95,109)(H,96,112)(H,97,116)(H,104,105)(H4,80,81,83)/t51-,52-,53-,54-,55-,56-,57-,58?,59-,60-,61-,64-/m0/s1

InChI key

WHNFPRLDDSXQCL-PDEGJNOBSA-N

General description

Synthetic α-melanocyte-stimulating hormone. Peptide hormone secreted by the anterior pituitary gland. Controls the intensity of pigmentation. A potent inhibitor of ICAM-1 expression in malignant melanocytes.

Packaging

Yes

Physical form

Supplied as trifluoroacetate salt. Sold on the basis of peptide content.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 2 months at -20°C.

Other Notes

Ac-Ser-Tyr-Ser-Met-Glu-His-Phe-Arg-Trp-Gly-Lys-Pro-Val-NH₂
Morandini, R., et al. 1998. J. Cell Physiol. 175, 276.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M4135M8764M8693
form

lyophilized

form

powder

form

powder

form

lyophilized solid

assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

200

manufacturer/tradename

Calbiochem®

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

water: 1 mg/mL

solubility

water: 1.00-1.04 mg/mL, clear, colorless

solubility

0.1% aqueous TFA: 1.00-1.04 mg/mL, clear, colorless

solubility

H2O: soluble 5 mg/mL


Still not finding the right product?

Explore all of our products under α-Melanocyte-Stimulating Hormone


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters