Skip to Content
Merck
All Photos(1)

Key Documents

05-1328

Sigma-Aldrich

Anti-PHD3 Antibody, clone 188e

Synonym(s):

EGL nine (C.elegans) homolog 3, HIF prolyl hydroxylase 3, HIF-prolyl hydroxylase 3, Hypoxia-inducible factor prolyl hydroxylase 3, Prolyl hydroxylase domain-containing protein 3, egl nine homolog 3, egl nine homolog 3 (C. elegans), egl nine-like protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Pricing and availability is not currently available.

antibody product type

primary antibodies

Quality Level

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU903961EMU147591EHU031881
esiRNA cDNA target sequence

CACGAACCCCAGACCTGTTTGTATCATCCGGGCTCCTTCCGGGCAGAAACAACTGAAAATGCACTTCAGACCCACTTATTTCTGCCACATCTGAGTCGGCCTGAGATAGACTTTTCCCTCTAAACTGGGAGAATATCACAGTGGTTTTTGTTAGCAGAAAATGCACTCCAGCCTCTGTACTCATCTAAGCTGCTTATTTTTGATATTTGTGTCAGTCTGTAAATGGATACTTCACTTTAATAACTGTTGCTTAGTAATTGGCTTTGTAGAGAAGCTGGAAAAAAATGGTTTTGTCTTCAACTCCTTTGCATGCCAGG

esiRNA cDNA target sequence

GCGTGCGCACCTTCCTGTCCTAGGCCAGGTGCCATGGCCGGCCAGGTGGGCTGCAGAGTGGGCTCCCTGCCCCTCTCTGCCTGTTCTGGACTGTGTTCTGGGCCTGCTGAGGATGGCAGAGCTGGTGTCCATCCAGCACTGACCAGCCCTGATTCCCCGACCACCGCCCAGGGTGGAGAAGGAGGCCCTTGCTTGGCGTGGGGGATGGCTTAACTGTACCTGTTTGGATGCTTCTGAATAGAAATAAAGTGGGTTTTCCCTGGAGGT

esiRNA cDNA target sequence

GAAGGCTGCGGATCAACAATCAGGCGCCTCTGCTTCCAGGGCGCCGGGGGCCTGACTCGGTGGTGAGTGCGGCTGCCTTCGTCAGGACGGGCGAGCGTGGCTCTCGACGGCTGGGCGGGCTAGCTGACTCCTCCTTCGGCCCGGAAGGCAGTTCTCAGGGCCGCCCGACCCCCGCTCCCTGCCTAAGTCCTTGGGCTTGGACTTGTATAACAGTTTTGCTTTCTTTTCCTTTCGGTTTATTTTTTCAGTCAACCCAGGCTAGTCTCGAATTTGCGGCAATCCTCCTGCCTCCAATCGTTCTAGGTGCTGGGATTACTGGTGTGCAGCACCTCGGCTGTCTCTTCAGATTTTCTGCAGGTT

esiRNA cDNA target sequence

TGGGAGTGACAGATGATGGAGAAGGAAGTCATATTCTTCAATCTCCATCAGCCAATGTGCTTCCAACCCTTCCTTTCCACGTCCTTCGTAGCTTGTTTAGCACTACACCTTTGACAACTGATGATGGTGTACTTCTAAGGCGGATGGCATTGGAAATTGGAGCCTTACACCTCATTCTTGTCTGTCTCTCTGCTTTGAGCCACCATTCCCCACGAGTTCCAAACTCTAGCGTGAATCAAACTGAGCCACAGGTGTCAAGCTCTCATAACCCTACATCAACAGAAGAACAACAGTTATATTGGGCCAAAGGGACTGGCTTTGGAACAGGCTCTACAGCTTCTGGGTGGGATGTGGAACAAGCCTTAACTAAGCAAAGGCTGGAAGAGGAACATGTTACCTGCCTTCTGCAGGTT

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized, lyophilized powder

form

lyophilized, lyophilized powder

form

lyophilized powder

form

lyophilized powder

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service