Skip to Content
Merck
All Photos(1)

Documents

03-189

Sigma-Aldrich

CBP (catalytic domain) human recombinant

CBP (catalytic domain) human recombinant protein expressed in E. Coli.

Synonym(s):

Human CBP (catalytic domain) protein, Human recombinant CBP, Upstate® human recombinant CBP (catalytic domain)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

biological source

human

Quality Level

Assay

>65% (SDS-PAGE)

mol wt

~48 kDa

manufacturer/tradename

Upstate®

technique(s)

enzyme immunoassay: suitable

UniProt accession no.

General description

Although originally termed histone acetyltransferases (HATs) for their lysine acetylation activity on histone N-terminal tails, p300 and its paralogue, CBP, have been shown to acetylate a variety of non-histone proteins including p53, DNA polymerase ß and nuclear import factors. p300/CBP acetylations play regulatory roles in transcription, DNA repair and replication, the cell cycle, p53 turnover, and nuclear import. In addition to functions which overlap with those of CBP, p300 has a number of unique functions, including a role in the induction of p21Waf1/Cip1 cell cycle inhibitor.

Source:
Catalytic domain (residues 1319-1710) of human CBP histone acetyltransferase

Specificity

[Varies with lot. For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.] XXX 250 pmol/min/µg assayed as production of CoA-SH from AcCoA in the presence of a peptide comprising human p53 residues 368-386. CoA-SH is determined colorimetrically by reaction with DTNB (5,5′-Dithiobis (2-nitrobenzoic acid).

Application

Acetyltransferase assays
CBP (catalytic domain) human recombinant protein expressed in E. Coli.

Physical form

Format: Purified
In liquid containing 50 mM Tris/HCl, pH 8.0, 0.1 mM EDTA, 10% glycerol. Frozen solution.

Storage and Stability

Stable for 12 months at -80°C from date of receipt. Avoid multiple freeze/thaw cycles.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service