Skip to Content

03-186

HDAC8 human recombinant

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.84
eCl@ss:
32160405
Technique(s):
enzyme immunoassay: suitable
Species reactivity:
-
Application:
EIA
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

assay

>50% (SDS-PAGE)

technique(s)

enzyme immunoassay: suitable

General description

HDAC8 is phosphorylated in vivo by PKA, resulting in decreased activity and hyperacetylation of HDAC8 targets, such as histones H3 and H4. In addition, HDAC8 can also be inhibited by trichostatin A. HDAC8 has been implicated in acute myeloid leukemia.
Human recombinant HDAC8 expressed in E. Coli
~42 kDa

Application

HDAC deacetylase assays

Physical form

In liquid containing 10 mM Tris, pH 7.5, 100 mM NaCl, 3 mM MgCl2 and 10% glycerol. Frozen solution.

Unit Definition: 1U = 1 pmol/min at 37°C using a standard flourometric activity assay to measure deacetylation activity

Preparation Note

Store at -80°C. The enzyme is stable on ice for 30 to 60 min, but may lose activity with dilution and/or prolonged storage on ice. It is recommended that thawing and addition of the enzyme to assay wells be done within as short a time as possible before start of the assay and that the thawed enzyme be kept on ice throughout. Best performance will be obtained by adding the chilled, undiluted enzyme directly to pre-warmed buffer and proceeding immediately to the addition of pre-warmed substrate. The remaining, unused enzyme should be refrozen quickly by, for example, snap freezing in a dry/ice ethanol bath or liquid nitrogen. The enzyme is stable to at least 4 freeze/thaw cycles. To minimize the number of freeze/thaw cycles, aliquot the HDAC8 into separate tubes before storing at -80°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library