KEY465196741
Key Organics / BIONET
methyl 2-[(2Z)-2-amino-2-[(phenylformamido)imino]ethoxy]benzoate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 2-{[(2Z)-2-amino-2-(benzoylhydrazono)ethyl]oxy}benzoate
Inchi Code
1S/C17H17N3O4/c1-23-17(22)13-9-5-6-10-14(13)24-11-15(18)19-20-16(21)12-7-3-2-4-8-12/h2-11,19H,18H2,1H3,(H,20,21)
InChI key
QKRKOEACIIKYAY-UHFFFAOYSA-N
Purity
90%
Country of Origin
GB
Physical Form
Solid
Product Link
https://www.keyorganics.net/sku/5F-010
Signal Word
Warning
Hazard Codes
MSDS
https://www.keyorganics.net/sds/5F-010
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.