Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249D6DD

J & W Pharmlab LLC

8-Bromo-6-chloro-3,4-dihydro-2H-benzo[1,4]oxazine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
284.97
MDL number:

Pricing and availability is not currently available.

Iupac Name

8-bromo-6-chloro-3,4-dihydro-2H-1,4-benzoxazine hydrochloride

Inchi Code

1S/C8H7BrClNO.ClH/c9-6-3-5(10)4-7-8(6)12-2-1-11-7;/h3-4,11H,1-2H2;1H

InChI key

XVTZJIQRUMCLKF-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

White Solid

Product Link

https://www.jwpharmlab.com/home/product/53R0087S

MSDS

https://www.jwpharmlab.com/home/product/53R0087S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library