Skip to Content
Merck
All Photos(1)

Key Documents

ENAH30490DA8

Enamine

[1-bromo-2-(2-methoxyethoxy)propan-2-yl]benzene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
273.17

Iupac Name

(1-bromo-2-(2-methoxyethoxy)propan-2-yl)benzene

Inchi Code

1S/C12H17BrO2/c1-12(10-13,15-9-8-14-2)11-6-4-3-5-7-11/h3-7H,8-10H2,1-2H3

InChI key

WCQKRYGKAXFSNO-UHFFFAOYSA-N

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-1134988

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library