Skip to Content
Merck
All Photos(1)

Key Documents

ENA429359211

Enamine

phenylmethanesulfinamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
155.22
MDL number:

mp

124-128

Iupac Name

phenylmethanesulfinamide

Inchi Code

1S/C7H9NOS/c8-10(9)6-7-4-2-1-3-5-7/h1-5H,6,8H2

InChI key

UKDZTPQQLLBYRG-UHFFFAOYSA-N

storage temp.

-10

Purity

95

Country of Origin

UA

Shipping Temperature

Ice Pack

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-187300

Melting Point

124-128

MSDS

https://www.enaminestore.com/catalog/EN300-187300


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library