Skip to Content
Merck

COMH93D65BFC

Combi-Blocks, Inc.

Methyl 4-(5-iodo-1,3-thiazol-2-yl)benzoate

Synonym(s):

methyl 4-(5-iodothiazol-2-yl)benzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H8O2N1S1I1
CAS Number:
Molecular Weight:
345.16
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H8INO2S/c1-15-11(14)8-4-2-7(3-5-8)10-13-6-9(12)16-10/h2-6H,1H3

Inchi Key

BAVRZQULDHRSHB-UHFFFAOYSA-N

Purity

98%

Country of Origin

CN





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library