Skip to Content
Merck
All Photos(1)

Key Documents

COMH04237483

Combi-Blocks, Inc.

8-Chloro-2-(3-chlorophenyl)quinoline-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
318.16
MDL number:

Iupac Name

8-chloro-2-(3-chlorophenyl)-4-quinolinecarboxylic acid

Inchi Code

1S/C16H9Cl2NO2/c17-10-4-1-3-9(7-10)14-8-12(16(20)21)11-5-2-6-13(18)15(11)19-14/h1-8H,(H,20,21)

InChI key

IVDWLWGPHBNIRA-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0981

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QZ-0981


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library