Skip to Content
Merck

COM448686243

Combi-Blocks, Inc.

Fmoc-Cit-OMe

Synonym(s):

methyl (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C22H25O5N3
CAS Number:
Molecular Weight:
411.46
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C22H25N3O5/c1-29-20(26)19(11-6-12-24-21(23)27)25-22(28)30-13-18-16-9-4-2-7-14(16)15-8-3-5-10-17(15)18/h2-5,7-10,18-19H,6,11-13H2,1H3,(H,25,28)(H3,23,24,27)/t19-/m0/s1

Inchi Key

CNUDAIIEDFQCCQ-IBGZPJMESA-N

Purity

96%

Storage temp.

Refrigerated

Country of Origin

CN



Still not finding the right product?

Explore all of our products under Fmoc-Cit-OMe




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library