Skip to Content
Merck

COM448684879

Combi-Blocks, Inc.

4-(2,4-Dimethoxyphenyl)benzoic acid

Synonym(s):

2',4'-dimethoxy[1,1'-biphenyl]-4-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H14O4
CAS Number:
Molecular Weight:
258.27
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H14O4/c1-18-12-7-8-13(14(9-12)19-2)10-3-5-11(6-4-10)15(16)17/h3-9H,1-2H3,(H,16,17)

Inchi Key

XVVFNXGUNVVYOR-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library