Skip to Content
Merck
All Photos(1)

Key Documents

COM448654073

Combi-Blocks, Inc.

2-Bromo-3,3,4,4,4-pentafluoro-1-butene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
224.96
MDL number:
Pricing and availability is not currently available.

Iupac Name

2-bromo-3,3,4,4,4-pentafluoro-1-butene

Inchi Code

1S/C4H2BrF5/c1-2(5)3(6,7)4(8,9)10/h1H2

InChI key

AAYAPGNGKQASNS-UHFFFAOYSA-N

Purity

95%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?QC-0456

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?QC-0456


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library