Skip to Content
Merck

COM448622207

Combi-Blocks, Inc.

6,7-Difluoro-1-methyl-1,3-benzimidazole

Synonym(s):

6,7-difluoro-1-methyl-1H-benzimidazole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H6N2F2
Molecular Weight:
168.15
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H6F2N2/c1-12-4-11-6-3-2-5(9)7(10)8(6)12/h2-4H,1H3

Inchi Key

PFYZSWKRVVPXTN-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library