Skip to Content
Merck

COM448614938

Combi-Blocks, Inc.

5-Chloro-2-methoxy-6-methylpyridine-3-boronic acid

Synonym(s):

(5-chloro-2-methoxy-6-methylpyridin-3-yl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H9O3N1Cl1B1
CAS Number:
Molecular Weight:
201.42
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H9BClNO3/c1-4-6(9)3-5(8(11)12)7(10-4)13-2/h3,11-12H,1-2H3

Inchi Key

CYQRZZIAQDQBHW-UHFFFAOYSA-N

Purity

98%

Storage temp.

Freezer

Country of Origin

CN



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library