C12H316CEB12
Chemcia Scientific, LLC
2-Amino-4-bromo-5-iodo-benzoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
methyl 2-amino-4-bromo-5-iodobenzoate
Inchi Code
1S/C8H7BrINO2/c1-13-8(12)4-2-6(10)5(9)3-7(4)11/h2-3H,11H2,1H3
InChI key
WSALXGZBDVNFLR-UHFFFAOYSA-N
storage temp.
0-5
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Brown Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=3291
MSDS
http://www.chemcia.com/product_fl02.asp?id=3291
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service