Skip to Content
Merck

Skip To

V-045

O-Desmethylvenlafaxine solution

1.0 mg/mL in methanol, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
10 700,00 CZK

About This Item

Empirical Formula (Hill Notation):
C16H25NO2
CAS Number:
Molecular Weight:
263.38
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:

10 700,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC=C(C(CN(C)C)C2(O)CCCCC2)C=C1

InChI

1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3

InChI key

KYYIDSXMWOZKMP-UHFFFAOYSA-N

Gene Information

General description

An analytical reference standard applicable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications such as urine drug testing, forensic analysis, or clinical toxicology. Also known as desvenlafaxine, O-desmethylvenlafaxine is an SNRI antidepressant marketed under the trade name Pristiq for the treatment of depression. O-desmethylvenlafaxine is also a major urinary metabolite of venlafaxine, an SNRI antidepressant sold as Effexor or Efexor and used to treat major depressive disorder, generalized anxiety disorder, and other anxiety disorders associated with depression.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V-007V-027D-906
description

1.0 mg/mL in methanol, certified reference material, Cerilliant®

description

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

description

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

description

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

format

single component solution

format

single component solution

format

single component solution

format

single component solution

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

concentration

1.0 mg/mL in methanol

concentration

100 μg/mL in methanol

concentration

100 μg/mL in methanol

concentration

1.0 mg/mL in methanol (as free base)

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL

packaging

ampule of 1 mL

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing

application(s)

clinical testing


Still not finding the right product?

Explore all of our products under O-Desmethylvenlafaxine solution


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup

target_organs

Eyes,Central nervous system


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters