Skip to Content
Merck

Skip To

L-044

Levorphanol tartrate solution

1.0 mg/mL in methanol (as free base), certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H23NO · C4H6O6
CAS Number:
Molecular Weight:
407.46
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland

concentration

1.0 mg/mL in methanol (as free base)

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC([C@](CCCC2)(CCN3C)[C@]2([H])[C@H]3C4)=C4C=C1.OC([C@H](O)[C@@H](O)C(O)=O)=O

InChI

1S/C17H23NO.C4H6O6/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17;5-1(3(7)8)2(6)4(9)10/h5-6,11,14,16,19H,2-4,7-10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t14-,16+,17+;1-,2-/m01/s1

InChI key

RWTWIZDKEIWLKQ-IWWMGODWSA-N

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M-236L-034B-037
format

single component solution

format

single component solution

format

single component solution

format

single component solution

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

drug control

estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland

drug control

-

drug control

-

drug control

-

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

Snap-N-Spike®/Snap-N-Shoot®

feature

(Snap-N-Spike®)

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®

manufacturer/tradename

Cerilliant®


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters