Skip to Content
Merck
All Photos(1)

Key Documents

H-119

Supelco

m-Hydroxycocaine solution

1.0 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
CZK 5,390.00

CZK 5,390.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
CZK 5,390.00

About This Item

Empirical Formula (Hill Notation):
C17H21NO5
CAS Number:
Molecular Weight:
319.35
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 5,390.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

N1([C@H]2C(C(CC1CC2)OC(=O)c3cc(ccc3)O)C(=O)OC)C

InChI

1S/C17H21NO5/c1-18-11-6-7-13(18)15(17(21)22-2)14(9-11)23-16(20)10-4-3-5-12(19)8-10/h3-5,8,11,13-15,19H,6-7,9H2,1-2H3/t11?,13-,14?,15?/m1/s1

InChI key

IQXBUEUAOWARGT-QZMFJGEPSA-N

General description

Meta-hydroxycocaine (m-hydroxycocaine) is a metabolite of cocaine. Measurement of m-hydroxycocaine in biological samples is indicative of cocaine use. This certified solution standard is suitable for use as starting material in calibrators and controls for GC/MS or LC/MS cocaine testing methods in forensic analysis, clinical toxicology, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service