Skip to Content
Merck

Skip To

C-256

Cannabichromevarinic acid (CBCVA) solution

1 mg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile), certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

5-Hydroxy-2-methyl-2-(4-methyl-3-penten-1-yl)-7-propyl-2H-1-benzopyran-6-carboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mL
Please contact Customer Service for Availability
7 080,00 CZK

About This Item

Empirical Formula (Hill Notation):
C20H26O4
CAS Number:
Molecular Weight:
330.42
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:

7 080,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL (1% DIPEA and 0.05% Ascorbic Acid in Acetonitrile)

format

single component solution

storage temp.

-10 to -25°C

General description

Snap-N-Spike®/Snap-N-Shoot®
This certified Snap-N-Spike® reference solution is suitable for numerous GC/MS or LC/MS applications from pharmaceutical testing and research to clinical toxicology or forensic analysis. Cannabichromevarinic acid (CBCVA) is a cannabinoid found in cannabis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F

flash_point_c

2.0 °C



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters