Sign In to View Organizational & Contract Pricing.
About This Item
Empirical Formula (Hill Notation):
C1013C6H11ClK2N2O4
Molecular Weight:
414.88
UNSPSC Code:
41116107
grade
certified reference material
feature
Snap-N-Spike®/Snap-N-Shoot®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol ((as clorazepate))
format
single component solution
storage temp.
−20°C
General description
This stable-labeled internal standard is suitable for quantification of clorazepate by LC/MS or GC/MS for clinical toxicology, forensic analysis, or urine drug testing applications. Clorazepate dipotassium is a long-acting benzodiazepine with anxiolytic, anticonvulsant, sedative and hypnotic properties. Clorazepate dipotassium, marketed under the brand name Tranxene, is a prodrug for nordiazepam, which is produced as an active metabolite.
Legal Information
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes,Central nervous system
Storage Class
3 - Flammable liquids
Flash Point (°F)
49.5 °F - closed cup
Flash Point (°C)
9.7 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
67-56-1
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


