Skip to Content
Merck

Skip To

C-203

Carfentanil-13C6 oxalate solution

100 μg/mL in methanol (as free base), certified reference material, ampule of 0.5 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
13C6C18H30N2O3 · C2H2O4
Molecular Weight:
490.50
UNSPSC Code:
41116107


grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 0.5 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

General description

This stable labeled internal standard is suitable for quantitation of carfentanil in fentanyl GC/MS or LC/MS methods for applications in clinical toxicology, forensic analysis, urine drug testing, pain prescription monitoring or isotope dilution.
Marketed under the trade name Wildnil as a general anesthetic for large animals, carfentanil hit the illicit market in 2016 as a new NPS drug or in combination with heroin. Carfentanil, also known as 4-carbomethoxyfentanyl, is a fentanyl analog 10,000 times more potent than morphine, making it one of the most potent synthetic opioids available.
This product will be shipped in a can containing one ampoule of C-203-0.5ML due to the compound′s acute toxicity.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class

3 - Flammable liquids

Flash Point (°F)

49.5 °F - closed cup

Flash Point (°C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

67-56-1

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters