Skip to Content
Merck

A-214

Adinazolam solution

1 mg/mL in acetonitrile, certified reference material, ampule of 1 mL, Cerilliant®

Synonym(s):

8-Chloro-N,N-dimethyl-6-phenyl-4H-[1,2,4]triazolo[4,3-a][1,4]benzodiazepine-1-methanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H18ClN5
CAS Number:
Molecular Weight:
351.83
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1 mg/mL in acetonitrile

format

single component solution

storage temp.

-10 to -25°C

SMILES string

Clc1cc2c(cc1)[n]3c(nnc3CN(C)C)CN=C2c4ccccc4

InChI

1S/C19H18ClN5/c1-24(2)12-18-23-22-17-11-21-19(13-6-4-3-5-7-13)15-10-14(20)8-9-16(15)25(17)18/h3-10H,11-12H2,1-2H3

InChI key

GJSLOMWRLALDCT-UHFFFAOYSA-N

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library