Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mL | Please contact Customer Service for Availability | 4 530,00 CZK |
About This Item
grade
certified reference material
Quality Segment
form
liquid
feature
SNAP-N-SPIKE®, SNAP-N-SHOOT®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
100 μg/mL in methanol (as free base)
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
Cl.[2H]c1c([2H])c([2H])c(c(O[C@H](CCNC)c2ccccc2)c1[2H])C([2H])([2H])[2H]
InChI
1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1/i1D3,6D,7D,8D,11D;
InChI key
LUCXVPAZUDVVBT-OJPCNXEKSA-N
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant® | description 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant® | description 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant® | description 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant® |
| application(s) forensics and toxicology | application(s) clinical testing | application(s) clinical testing | application(s) forensics and toxicology |
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| form liquid | form liquid | form liquid | form liquid |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. −20°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C |
signalword
Danger
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
target_organs
Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
49.5 °F - closed cup
flash_point_c
9.7 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
67-56-1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





