Skip to Content
Merck
All Photos(1)

Key Documents

880510P

Avanti

14:0 PEG550 PE

Avanti Research - A Croda Brand 880510P, powder

Synonym(s):

1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-550] (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CZK 7,410.00

CZK 7,410.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
CZK 7,410.00

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25

CZK 7,410.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880510P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880510P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C59H116NO22P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-57(61)79-54-56(82-58(62)27-25-23-21-19-17-15-13-11-9-7-5-2)55-81-83(64,65)80-29-28-60-59(63)78-53-52-77-51-50-76-49-48-75-47-46-74-45-44-73-43-42-72-41-40-71-39-38-70-37-36-69-35-34-68-33-32-67-31-30

InChI key

KEIXSZLRQVRTCP-DRZXTAICSA-N

General description

14:0 PEG550 PE is a synthetic anionic lipid. It contains 14-carbon acyl chains conjugated to phosphoethanolamine headgroup and polyethylene-glycol.[1] The molecular weight and exact mass are averages based on the polydispersity of PEG.[2]

Application

14:0 PEG550 PE might be used as lipid additive to study its effect on the phase properties of hydrated monoolein.[1] It is also suitable for use in the formation of liposomes encapsulating hemoglobin (LEHs).[3]

Biochem/physiol Actions

14:0 PEG550 PE is applicable in targeted drug delivery. It favors the formation of hexagonal (HI)-phase lipids.[1]

Packaging

5 mL Clear Glass Sealed Ampule (880510P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vadim Cherezov et al.
Biophysical journal, 83(6), 3393-3407 (2002-12-24)
Hydrated monoolein forms the cubic-Pn3m mesophase that has been used for in meso crystallization of membrane proteins. The crystals have subsequently provided high-resolution structures by crystallographic means. It is possible that the hosting cubic phase created by monoolein alone, which
Dian R Arifin et al.
Artificial cells, blood substitutes, and immobilization biotechnology, 33(2), 137-162 (2005-06-18)
Liposomes encapsulating hemoglobin (LEHs) surface-conjugated with 2000 and 550 Da poly(ethylene glycol) (PEG) were produced via extrusion through 400, 200 and 100 nm pore diameter membranes in two types of phosphate buffer with different ionic strengths. The lipid bilayers were
Brian J Nablo et al.
Biophysical journal, 95(3), 1157-1164 (2008-07-23)
Nonelectrolyte polymers of poly(ethylene glycol) (PEG) were used to estimate the diameter of the ion channel formed by the Bacillus anthracis protective antigen 63 (PA(63)). Based on the ability of different molecular weight PEGs to partition into the pore and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service