Skip to Content
Merck
All Photos(1)

Key Documents

880280P

Avanti

C16 PEG5000 Ceramide

Avanti Research - A Croda Brand 880280P, powder

Synonym(s):

N-palmitoyl-sphingosine-1-{succinyl[methoxy(polyethylene glycol)5000]}

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
CZK 12,400.00

CZK 12,400.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
CZK 12,400.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CZK 12,400.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 25 mg (880280P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880280P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCC)=O)COC(CCC([(OCH2CH2)113OCH3])=O)=O

General description

Ceramides are lipid molecules of stratum corneum. They belong to the group of sphingolipids.[1] C16 PEG5000 Ceramide is categorized as polymers & polymerizable lipids.

Application

C16 PEG5000 Ceramide might be used to stabilize nanosized bilayer disks[2] and in the preparation of lipid aggregates.[3]

Biochem/physiol Actions

Topical applications containing ceramides is known to repair and replenish the lipid levels in stratum corneum.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (880280P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The design and development of anti-aging formulations
Skin Aging Handbook, 291-325 (2009)
Nanosized bilayer disks: attractive model membranes for drug partition studies
Johansson E, et al.
Biochimica et Biophysica Acta - Biomembranes, 1768(6), 1518-1525 (2007)
Melittin-Lipid interaction: A comparative study using liposomes, micelles and bilayerdisks
Lundquist A, et al.
Biochimica et Biophysica Acta - Biomembranes, 1778(10), 2210-2216 (2008)
Hudson P Pace et al.
Analytical chemistry, 90(21), 13065-13072 (2018-10-24)
Over the last two decades, supported lipid bilayers (SLBs) have been extensively used as model systems to study cell membrane structure and function. While SLBs have been traditionally produced from simple lipid mixtures, there has been a recent surge in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service