Skip to Content
Merck
All Photos(1)

Key Documents

878113C

Avanti

C16-20:4 PC

Avanti Research - A Croda Brand 878113C

Synonym(s):

1-O-hexadecyl-2-arachidonoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 12,100.00

CZK 12,100.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CZK 12,100.00

About This Item

Empirical Formula (Hill Notation):
C44H82NO7P
CAS Number:
Molecular Weight:
768.10
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CZK 12,100.00


Please contact Customer Service for Availability

Request a Bulk Order

form

liquid

packaging

pkg of 1 × 1 mL (878113C-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 878113C

concentration

5 mg/mL (878113C-5mg)

lipid type

bioactive lipids
phosphoglycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C44H82NO7P/c1-6-8-10-12-14-16-18-20-22-23-24-25-27-29-31-33-35-37-44(46)52-43(42-51-53(47,48)50-40-38-45(3,4)5)41-49-39-36-34-32-30-28-26-21-19-17-15-13-11-9-7-2/h14,16,20,22,24-25,29,31,43H,6-13,15,17-19,21,23,26-28,30,32-42H2,1-5H3/b16-14-,22-20-,25-24-,31-29-/t43-/m1/s1

InChI key

DUUSFCFZBREELS-WWBBCYQPSA-N

General description

C16-20:4 PC is an analog to platelet-activation factor (PAF).

Packaging

5 mL Clear Glass Sealed Ampule (878113C-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service