Skip to Content
Merck
All Photos(1)

Key Documents

870740P

Avanti

16:0 Ether Coenzyme A

Avanti Research - A Croda Brand 870740P, powder

Synonym(s):

hexadecyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
CZK 10,200.00

CZK 10,200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
CZK 10,200.00

About This Item

Empirical Formula (Hill Notation):
C37H77N10O16P3S
CAS Number:
Molecular Weight:
1043.05
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CZK 10,200.00


Please contact Customer Service for Availability

Request a Bulk Order

form

powder

packaging

pkg of 1 × 5 mg (870740P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870740P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSCCCCCCCCCCCCCCCC)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C37H68N7O16P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21-64-22-20-39-28(45)18-19-40-35(48)32(47)37(2,3)24-57-63(54,55)60-62(52,53)56-23-27-31(59-61(49,50)51)30(46)36(58-27)44-26-43-29-33(38)41-25-42-34(29)44;;;/h25-27,30-32,36,46-47H,4-24H2,1-3H3,

InChI key

XESJXIGUEOLPCN-PJGFIIEVSA-N

General description

16:0 Ether Coenzyme A, also known as hexadecyl-CoA, is a thioether analog of palmitoyl-CoA. It has an ability to inhibit adipose triglyceride lipase (ATGL).[1]

Application

16:0 Ether Coenzyme A has been used as an inhibitor of adipose triglyceride lipase (ATGL).[1]

Packaging

5 mL Amber Glass Screw Cap Vial (870740P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Harald M Nagy et al.
Biochimica et biophysica acta, 1841(4), 588-594 (2014-01-21)
Adipose triglyceride lipase (ATGL) is required for efficient mobilization of triglyceride (TG) stores in adipose tissue and non-adipose tissues. Therefore, ATGL strongly determines the availability of fatty acids for metabolic reactions. ATGL activity is regulated by a complex network of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service