Skip to Content
Merck
All Photos(1)

Documents

860695P

Avanti

Glucosyl(β) Sphingosine-d7

Avanti Research - A Croda Brand (860695P), powder

Synonym(s):

D-glucosyl-β-1,1’-D-erythro-sphingosine-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40D7NO7
CAS Number:
Molecular Weight:
468.68
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860695P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand (860695P)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)[C@](N)([H])CO[C@H](O1)[C@H](O)[C@@H](O)[C@@H]([C@H]1CO)O

General description

Glucosyl(β) Sphingosine-d7 is a deuterated derivative of glucosyl(β) sphingosine.

Biochem/physiol Actions

Glucosyl sphingosine acts as a biomarker for Gaucher′s disease.

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glucosylsphingosine is a highly sensitive and specific biomarker for primary diagnostic and follow-up monitoring in Gaucher disease in a non-Jewish, Caucasian cohort of Gaucher disease patients
Rolfs A, et al.
Testing, 8(11) (2013)
Glucosylsphingosine is a highly sensitive and specific biomarker for primary diagnostic and follow-up monitoring in Gaucher disease in a non-Jewish, Caucasian cohort of Gaucher disease patients
Rolfs A, et al.
PLoS ONE, 8(11) (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service