Skip to Content
Merck
All Photos(2)

Key Documents

860397P

Avanti

16:0-d31 Lyso PC

Avanti Research - A Croda Brand 860397P, powder

Synonym(s):

1-palmitoyl-d31-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CZK 28,600.00

CZK 28,600.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
CZK 28,600.00

About This Item

Empirical Formula (Hill Notation):
C24H19NO7PD31
CAS Number:
Molecular Weight:
526.82
UNSPSC Code:
12352100
NACRES:
NA.25

CZK 28,600.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 10 mg (860397P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860397P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COC(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O

InChI

1S/C24H50NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-24(27)30-21-23(26)22-32-33(28,29)31-20-19-25(2,3)4/h23,26H,5-22H2,1-4H3/t23-/m1/s1/i1D3,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,14D2,15D2,16D2,17D2,18D2

InChI key

ASWBNKHCZGQVJV-NMMBEXRYSA-N

General description

16:0-d31 Lyso PC is a deuterated lysophosphatidylcholine (lyso pc) in which 31 protons of palmitoyl are replaced by deuterium.
Deuterated fatty acids experience exchange of the deuteriums on the alpha carbon to the carbonyl, i.e., C2 position, and will therefore be a mixture of compounds that are fully deuterated and partially deuterated at that position.

Application

16:0-d31 Lyso PC has been used to measure acyltransferase activity in lysophospholipid acyltransferase (LPLAT) assays.[1]

Packaging

5 mL Amber Glass Screw Cap Vial (860397P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidylcholine acyltransferase 3 is the key enzyme for incorporating arachidonic acid into glycerophospholipids during adipocyte differentiation
Eto M, et al.
International Journal of Molecular Sciences, 13(12), 16267-16280 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service