Skip to Content
Merck
All Photos(2)

Documents

860353P

Avanti

16:0 PC-d13

Avanti Research - A Croda Brand 860353P, powder

Synonym(s):

1,2-dipalmitoyl-sn-glycero-3-phosphocholine-1,1,2,2-d4-N,N,N-trimethyl-d9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H67NO8PD13
CAS Number:
Molecular Weight:
747.12
UNSPSC Code:
12352100
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 10 mg (860353P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860353P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OC([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O

General description

16:0 PC-d13 is a deuterated phosphocholine (PC) in which 13 protons of dipalmitoyl are replaced by deuterium.

Application

16:0 PC-d13 has been used in the preparation of asymmetric large unilamellar vesicles (aLUVs) to measure lipid translocation.

Packaging

5 mL Amber Glass Screw Cap Vial (860353P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1H NMR shows slow phospholipid flip-flop in gel and fluid bilayers
Marquardt D, et al.
Langmuir, 33(15) (2017)
Drew Marquardt et al.
Langmuir : the ACS journal of surfaces and colloids, 33(15), 3731-3741 (2017-01-21)
We measured the transbilayer diffusion of 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) in large unilamellar vesicles, in both the gel (Lβ') and fluid (Lα) phases. The choline resonance of headgroup-protiated DPPC exchanged into the outer leaflet of headgroup-deuterated DPPC-d13 vesicles was monitored using 1H

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service