Skip to Content
Merck
All Photos(1)

Documents

855773C

Avanti

2-18:1 Lyso PC

Avanti Research - A Croda Brand, chloroform solution

Synonym(s):

1-Hydroxy-2-oleoyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H52NO7P
CAS Number:
Molecular Weight:
521.67
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

chloroform solution

packaging

pkg of 1 × 1 mL (855773C-10MG)
pkg of 1 × 1 mL (855773C-1MG)
pkg of 1 × 1 mL (855773C-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

1 mg/mL in chloroform (855773C-1MG)
10 mg/mL in chloroform (855773C-10MG)
5 mg/mL in chloroform (855773C-5MG)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\CCCCCCCC)=O)CO)=O

InChI

1S/C26H52NO7P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-26(29)34-25(23-28)24-33-35(30,31)32-22-21-27(2,3)4/h12-13,25,28H,5-11,14-24H2,1-4H3/b13-12-/t25-/m1/s1

InChI key

SULIDBRAXVDKBU-PTGWMXDISA-N

Application

2-18:1 Lyso PC has been used in the preparation of multilamellar lipid vesicles (MLVs).

Packaging

5 mL Clear Glass Sealed Ampule (855773C-1MG, 855773C-5MG, and 855773C-10MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pingan Chang et al.
Biomolecules, 9(12) (2019-12-15)
: Neuropathy target esterase (NTE) is an endoplasmic reticulum (ER)-localized phospholipase that deacylates phosphatidylcholine (PC) and lysophosphatidylcholine (LPC). Loss-of-function mutations in the human NTE gene have been associated with a spectrum of neurodegenerative disorders such as hereditary spastic paraplegia, ataxia
Rapid affinity chromatographic isolation method for LDL in human plasma by immobilized chondroitin-6-sulfate and anti-apoB-100 antibody monolithic disks in tandem
Liangsupree T, et al.
Scientific Reports, 9(1), 1-10 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service