Skip to Content
Merck
All Photos(2)

Documents

850555O

Avanti

(2S,3S)-Phytantriol

Avanti Research - A Croda Brand 850555O

Synonym(s):

(2S,3S)-3,7,11,15-tetramethylhexadecane-1,2,3-triol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H42O3
CAS Number:
Molecular Weight:
330.55
UNSPSC Code:
12161902
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 25 mg (850555O-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850555O

shipped in

dry ice

storage temp.

−20°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCC[C@](C)(O)[C@@H](O)CO

InChI

1S/C20H42O3/c1-16(2)9-6-10-17(3)11-7-12-18(4)13-8-14-20(5,23)19(22)15-21/h16-19,21-23H,6-15H2,1-5H3

InChI key

CGIHFIDULQUVJG-UHFFFAOYSA-N

General description

Phytantriol (3,7,11,15-tetramethylhexadecane-1,2,3-triol) is a lipid, that is mainly used in cosmetics.
The temperature−composition phase diagram of phytantriol/water mixtures is qualitatively similar to that of aqueous glycerol monooleate so its stability makes it an interesting alternative to exploit various applications of the cubic liquid crystalline phases.

Biochem/physiol Actions

Phytantriol (PT; 3,7,11,15- tetramethylhexadecane-1,2,3-triol) do not hydrolyze. It possesses good structural stability and higher purity over glycerylmonooleate (GMO) or GMO-based derivatives. It helps to retain moisture in the skin and serves as a penetration promoter, hence can be used in transdermal drug delivery.

Packaging

5 mL Clear Glass Sealed Ampule (850555O-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Internally self-assembled submicrometer emulsions stabilized by spherical nanocolloids: finding the free nanoparticles in the aqueous continuous phase
Salonen A, et al.
Langmuir, 26(11), 7981-7987 (2010)
Impurities in commercial phytantriol significantly alter its lyotropic liquid-crystalline phase behavior
Dong YD, et al.
Langmuir, 24(13), 6998-7003 (2008)
S B Rizwan et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 79(1), 15-22 (2011-01-18)
Different delivery strategies to improve the immunogenicity of peptide/protein-based vaccines are currently under investigation. In this study, the preparation and physicochemical characterisation of cubosomes, a novel lipid-based particulate system currently being explored for vaccine delivery, was investigated. Cubosomes were prepared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service