Skip to Content
Merck
All Photos(2)

Documents

850359P

Avanti

16:1 (Δ9-Trans) PC

1,2-dipalmitelaidoyl-sn-glycero-3-phosphocholine, powder

Synonym(s):

1,2-di-(9E-hexadecenoyl)-sn-glycero-3-phosphocholine; PC(16:1(9E)/16:1(9E))

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H76NO8P
CAS Number:
Molecular Weight:
730.01
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (850359P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850359P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C/CCCCCC)=O)COC(CCCCCCC/C=C/CCCCCC)=O)=O

General description

16:1 (Δ9-Trans) PC (1,2-dipalmitelaidoyl-sn-glycero-3-phosphocholine) is a lipid derivative which has two 16 carbon fatty acid chains, each having one trans double bond at 9th carbon.
The list of Phosphatidylcholine products offered by Avanti is designed to provide compounds having a variety of physical properties. Products available include short chain (C3-C8 are water soluble and hygroscopic), saturated, multi-unsaturated and mixed acid PC′s. All of the products are purified by HPLC, and special precautions are taken to protect the products for oxidization and hydrolysis.

Application

16:1 (Δ9-Trans) PC (1,2-dipalmitelaidoyl-sn-glycero-3-phosphocholine) may be used in liposomes to study the cell proliferation of several cell lines and as a neutral or helper lipid to prepare liposomes.

Biochem/physiol Actions

Phosphatidylcholine (PC) helps to inhibit the penetrance of bacteria.

Packaging

5 mL Clear Glass Sealed Ampule (850359P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deciphering the functional composition of fusogenic liposomes
Kolavsinac R, et al
International Journal of Molecular Sciences (2018)
Interference of phosphatidylcholines with in-vitro cell proliferation?No flock without black sheep
Werlein A, et al.
BBA Clinical (2015)
Parian AM
Integrative Medicine (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service