Skip to Content
Merck
All Photos(2)

Key Documents

840503C

Avanti

18:0-18:1 PG

1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), chloroform

Synonym(s):

1-octadecanoyl-2-(9Z-octadecenoyl)-sn-glycero-3-phospho-(1′racglycerol) (sodium salt); SOPG; PG(18:0/18:1(9Z)); 110652

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80O10PNa
CAS Number:
Molecular Weight:
799.04
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840503C-25mg)
pkg of 2 × 4 mL (840503C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840503C

concentration

10 mg/mL (840503C-25mg)
25 mg/mL (840503C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/CCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C42H81O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-41(45)49-37-40(38-51-53(47,48)50-36-39(44)35-43)52-42(46)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h18,20,39-40,43-44H,3-17,19,21-38H2,1-2H3,(H,47,48);/q;+1/p-1/b20-18-;/t39?,40-;/m1./s1

InChI key

AZUBILBUHSANJX-VDVGXYCESA-M

General description

18:0-18:1 PG (SOPG/1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) is a hydrogenated phospholipid. It has two additional methylene groups in its saturated sn-1 chain. The thylakoid membranes of higher plants and cyanobacteria has phosphatidylglycerol (PG).

Application

18:0-18:1 PG (1-stearoyl-2-oleoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) may be used:
  • as a component in biocompatible phospholipid bilayer to coat the gold nanoparticles (GNPs) surface
  • in liposomes preparation
  • in the formation of lipid monolayer

Biochem/physiol Actions

Phosphatidylglycerol (PG) helps to maintain the stability of photosystem I and II protein complexes. It is involved in bacterial defense mechanisms.

Packaging

5 mL Clear Glass Sealed Ampule (840503C-200mg)
5 mL Clear Glass Sealed Ampule (840503C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianjun Pan et al.
Biochimica et biophysica acta, 1818(9), 2135-2148 (2012-05-16)
We have determined the molecular structures of commonly used phosphatidylglycerols (PGs) in the commonly accepted biologically relevant fluid phase. This was done by simultaneously analyzing small angle neutron and X-ray scattering data, with the constraint of measured lipid volumes. We
Meijing Wang et al.
Biochimica et biophysica acta, 1831(6), 1089-1097 (2013-02-06)
Gold nanoparticles (GNPs) have been applied as diagnostic and therapeutic agents because they can be targeted, localized, and be heated to cause cell death. However, their use has been limited by their relatively low biocompatibility. In this work, we coated
J P Michel et al.
Biochimica et biophysica acta, 1848(11 Pt A), 2967-2979 (2015-09-08)
Bacterial resistance to antibiotics has become today a major public health issue. In the development of new anti-infectious therapies, antimicrobial peptides appear as promising candidates. However, their mechanisms of action against bacterial membranes are still poorly understood. We describe for
J P Michel et al.
Langmuir : the ACS journal of surfaces and colloids, 33(41), 11028-11039 (2017-09-19)
The outer membrane (OM) of Gram-negative bacteria is a complex and asymmetric bilayer that antimicrobial peptides must disrupt in order to provoke the cell lysis. The inner and external leaflets of the OM are mainly composed of phospholipids (PL), and
Joury S van 't Klooster et al.
eLife, 9 (2020-04-18)
Yeast tolerates a low pH and high solvent concentrations. The permeability of the plasma membrane (PM) for small molecules is low and lateral diffusion of proteins is slow. These findings suggest a high degree of lipid order, which raises the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service