Skip to Content
Merck
All Photos(2)

Key Documents

810266P

Avanti

C11 TopFluor Galactosyl Ceramide

N-[11-(dipyrrometheneboron difluoride)undecanoyl]-D-galactosyl-β1-1′-D-erythro-sphingosine, powder

Synonym(s):

N-[11-(TopFluor)undecanoyl]-D-galactosyl-β1-1′-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
CZK 10,600.00
810266P-1MG
CZK 18,200.00

CZK 10,600.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 μG
CZK 10,600.00
810266P-1MG
CZK 18,200.00

About This Item

Empirical Formula (Hill Notation):
C48H80N3O8F2
CAS Number:
Molecular Weight:
865.16
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

CZK 10,600.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 250 μg (810266P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810266P

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

Galactosyl Ceramide is a glycosphingolipid, predominantly present in myelin sheath.[1]

Application

C11 TOPFLUOR Galactosyl Ceramide is suitable for in vivo labelling of antigen-presenting cells (APCs).[2]

Biochem/physiol Actions

Abnormal accumulation of galactosyl ceramide in the central nervous system, due to failure in its degradation process results in globoid cell leukodystrophy.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (810266P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Biosynthesis of Polysaccharides (2012)
Multiple Sclerosis: A Critical Conspectus, 195-195 (2012)
iNKT cell emigration out of the lung vasculature requires neutrophils and monocyte-derived dendritic cells in inflammation
Thanabalasuriar A, et al.
Testing, 16(12), 3260-3272 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service