Skip to Content
Merck
All Photos(1)

Key Documents

ATC311765492

Atlantic Research Chemicals Ltd

4-Chloropyridine-2-carbonyl chloride hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
176
MDL number:

Iupac Name

4-chloro-2-pyridinecarbonyl chloride

Inchi Code

1S/C6H3Cl2NO/c7-4-1-2-9-5(3-4)6(8)10/h1-3H

InChI key

FYBNFLRGZHGUDY-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=CC00204

MSDS

https://atlantic-chemicals.com/product/?code=CC00204


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library