Skip to Content
Merck
All Photos(1)

Key Documents

ATE514314668

AstaTech, Inc.

TERT-BUTYL 2-AMINO-1-(METHOXYMETHYL)ETHYLCARBAMATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
204.27
MDL number:

Iupac Name

tert-butyl 2-amino-1-(methoxymethyl)ethylcarbamate

Inchi Code

1S/C9H20N2O3/c1-9(2,3)14-8(12)11-7(5-10)6-13-4/h7H,5-6,10H2,1-4H3,(H,11,12)

InChI key

RIQNSCBRYGWVTD-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

95%

Shipping Temperature

Normal

Physical Form

Colorless to Yellow Sticky Oil to Semi-Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=79250

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=W10024


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library