Skip to Content
Merck
All Photos(1)

Documents

APO455836577

Apollo Scientific Ltd

2-Hydroxy-3-(trifluoromethyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
163.1
MDL number:

bp

150-155

Iupac Name

3-(trifluoromethyl)-2-pyridinol

Inchi Code

1S/C6H4F3NO/c7-6(8,9)4-2-1-3-10-5(4)11/h1-3H,(H,10,11)

InChI key

JHDCDEHVUADNKQ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid-Powder

Product Link

https://store.apolloscientific.co.uk/product/2-hydroxy-3-trifluoromethylpyridine

Synonym

3-(Trifluoromethyl)pyridin-2-ol, 3-(Trifluoromethyl)pyridin-2(1H)-one

Boiling point

150-155

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-hydroxy-3-trifluoromethylpyridine


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service