Skip to Content
Merck
All Photos(1)

Key Documents

APO455825732

Apollo Scientific Ltd

6-Amino-2,3-dichlorobenzonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
187.03
MDL number:

bp

141-144

Iupac Name

6-amino-2,3-dichlorobenzonitrile

Inchi Code

1S/C7H4Cl2N2/c8-5-1-2-6(11)4(3-10)7(5)9/h1-2H,11H2

InChI key

RYMBKSPMCKFIGN-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/6-amino-23-dichlorobenzonitrile

Boiling point

141-144

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/6-amino-23-dichlorobenzonitrile


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library