Z760323
Ohaus® MB-23 and MB-25 moisture analyzers
Model MB25, AC/DC input 110 V AC
Synonym(s):
moisture meters
About This Item
Recommended Products
manufacturer/tradename
Ohaus® MB25 US
weighing capacity measuring range
110 g
parameter
50-160 °C temp. limit
W × H × depth
17 cm × 13 cm × 28 cm
platform diam.
90 mm
Related Categories
General description
- easy to clean design
- heating chamber is low maintenance
- compact footprint
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)