Z743293
Julabo® adaptors
quad distributing, 1 1/4 inch female with barbed fittings, for 1 inch i.d. tubing, pack of 2
Synonym(s):
barbed adaptor
All Photos(1)
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
Recommended Products
packaging
pack of 2
manufacturer/tradename
Julabo 8970524
General description
For use with Julabo® FL Series Recirculating Cooler / Chillers , 2 Quad distributing adapters, G 1 1/4″ female, with barbed fittings for tubing 1″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. G 1¼” female thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for 1” ID tubing.
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)