Key Documents
Z743198
Julabo® bath test tube racks
to fit 90 test tubes, diam. 11 mm (12 mm), stainless steel
Synonym(s):
tube holder
About This Item
UNSPSC Code:
41103706
NACRES:
NB.77
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970346
parameter
100 °C max. temp.
diam.
11 mm (12 mm)
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.