Documents
Z742345
2MAG mixBATHcover for MIXdrive 6
polycarbonate
Synonym(s):
water bath lid
About This Item
UNSPSC Code:
41103706
NACRES:
NB.72
material
polycarbonate
manufacturer/tradename
2mag 40806
W × D × H
315 mm × 575 mm × 8 mm
weight
1.5 kg
General description
Chemically resistant, transparent cover for mixBATH, consisting of polycarbonate, for minimization of evaporation effects.
Different cutouts for the perfect positioning of the
thermostat in the case of using the stirring drives
MIXdrive 6 or MIXdrive 15 immersed into the mixBATH.