Skip to Content
Merck
All Photos(1)

Key Documents

Z742310

Rocking platform for RotoBot Mini

size 6 in. × 10 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
CZK 6,560.00

CZK 6,560.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EA
CZK 6,560.00

About This Item

UNSPSC Code:
41103811
NACRES:
NB.73

CZK 6,560.00


Please contact Customer Service for Availability

manufacturer/tradename

Benchmark Scientific

size

6 in. × 10 in.

Looking for similar products? Visit Product Comparison Guide

General description

For rocking applications, this platform with a non-slip rubber mat is an accessory for the Rotobot and Rotobot Mini rotating mixers. Once attached, simply select the "PP" mode to turn the rotator into a platform rocker.

Other Notes

For use with the Rotobot Rotating Mixer Z742300/1/2 (Benchmark Scientific number R4045).

Legal Information

Rotobot is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service