Key Documents
Z682748
Ace drying flask, pear-shaped, with side indents,
capacity 2000 mL, joint: ST/NS 24/40, plastic-coated glass
Synonym(s):
Aldrich® drying flask, pear-shaped, with side indents
Select a Size
1 EA
CZK 7,420.00
CZK 7,420.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 7,420.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
CZK 7,420.00
Please contact Customer Service for Availability
material
pear-shaped flask
plastic-coated glass
single-neck
Joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
case of 1
manufacturer/tradename
Ace Glass 3994-126
capacity
2000 mL
General description
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)