Z678031
Finnpipette® F2 multichannel pipette
volume 5-50 μL
Select a Size
CZK 35,000.00
Select a Size
About This Item
CZK 35,000.00
Recommended Products
feature
12-channel
color-coded yellow
electronic: no
packaging
pack of 1 ea
manufacturer/tradename
Finn 4662050
volume
5-50 μL
General description
- Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
- Big display with white numbers on black background for increased visibility
- Double action pipetting button with very light and smooth pipetting action
- Fully autoclavable
- Super blow-out piston with volumes of 50 μl and below
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)