Key Documents
Z677949
Finntip® Flex pipette tips
Finntip Flex 300, non-sterile
Select a Size
1000 EA
CZK 2,220.00
CZK 2,220.00
Please contact Customer Service for Availability
Select a Size
Change View
1000 EA
CZK 2,220.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
300 μL
volume range:
5-300 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 1000 ea (Bag)
manufacturer/tradename:
Finn 94060520
CZK 2,220.00
Please contact Customer Service for Availability
material
colorless tip
orange
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
manufacturer/tradename
Finn 94060520
maximum volume
300 μL
volume range
5-300 μL
General description
Finntip Flex pipette tips are easy-to-use, soft, flexible high-end tips. They are molded with diamond polished tooling for an extra smooth tip surface, which prevents liquid retention and achieves better pipetting accuracy.
Other Notes
Browse all pipette tips
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)