Skip to Content
Merck
All Photos(1)

Key Documents

Z673811

Hanna HI 2210 & HI 2211 bench pH meters

HI 2210, AC/DC input 110 V AC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603

manufacturer/tradename

Hanna HI 2210-01

availability

not available in EU

pH measuring range

-2.00-16.00, accuracy: ±0.01

temperature measuring range

-9.9-120.0 °C, accuracy: ±0.5 °C

Looking for similar products? Visit Product Comparison Guide

General description

  • Extra large LCD
  • Automatic Calibration
  • 5 Standard Buffers
  • Simultaneous display of pH and Temperature
  • Automatic Temperature Compensation
  • Stability indicator

Supplied with HI 1131B glass body pH electrode, HI 7662 temperature probe, electrode holder, buffer solutions pH 4.01 & 7.01, electrolyte solution, 12 V DC power adapter and instruction manual.

These units are compatible with a wide range of pH electrodes with BNC connectors.

For UK power adapter please order HI 710012 (Z673978)
The new microprocessor based HI 2211 and HI 2210 benchtop pH meters from HANNA are an affordable solution for laboratory testing without compromising accuracy. These instruments feature an extra large (6.5 in. x 1.5 in.) LCD which displays pH and temperature readings simultaneously. The large LCD also features a number of prompts and symbols that guide the user during the calibration operation, as well as during measurements.

  • Extra large LCD
  • Automatic Calibration
  • 5 Standard Buffers
  • Simultaneous display of pH and Temperature
  • Automatic Temperature Compensation
  • Stability indicator

Supplied with HI 1131B glass body pH electrode, HI 7662 temperature probe, electrode holder, buffer solutions pH 4.01 & 7.01, electrolyte solution, 12 V DC power adapter and instruction manual.

These units are compatible with a wide range of pH electrodes with BNC connectors.

For UK power adapter please order HI 710012 (Z673978)
mV - None

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service